Europium(III)trifluormethansulfonat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Europium(III) trifluoromethanesulphonate, Europium triflate

M-Clarity™ Qualitätsstufe = MQ100

425680-1G 425680-5G
SIAL425680-1GEA 57.3 EUR
SIAL425680-1G SIAL425680-5G
Europium(III)trifluormethansulfonat, Sigma-Aldrich®
Europium(III)trifluormethansulfonat
Formel: C₃EuF₉O₉S₃
Molecular Weight: 599,17 g/mol
MDL: MFCD00209587
CAS-Nummer: 52093-25-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR