(±)-α-Terpineol, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: α-Terpineol, Terpineol, 2-(4-Methylcyclohex-3-en-1-yl)propan-2-ol

M-Clarity™ Qualitätsstufe = MQ100

432628-250ML 432628-50ML
SIAL432628-250MLEA 265 EUR
SIAL432628-250ML SIAL432628-50ML
(±)-α-Terpineol, Sigma-Aldrich®
(±)-α-Terpineol
Formel: C₁₀H₁₈O
Molecular Weight: 154,25 g/mol
MDL: MFCD00001557
CAS-Nummer: 98-55-5
EINECS: 202-680-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR