Indium(III)trifluormethansulfonat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Indium(III) trifluoromethanesulphonate, Indium triflate
M-Clarity™ Qualitätsstufe = MQ100
442151-25G
442151-5G
SIAL442151-25GEA
368
EUR
SIAL442151-25G
SIAL442151-5G
Indium(III)trifluormethansulfonat, Sigma-Aldrich®
Indium(III)trifluormethansulfonat
Formel:
C₃F₉InO₉S₃ Molecular Weight: 562,03 g/mol |
MDL:
MFCD00144478 CAS-Nummer: 128008-30-0 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...