Diethylenglycolmonohexylether, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-(2-(Hexyloxy)ethoxy)ethanol, Diethylene glycol mono-n-hexyl ether

M-Clarity™ Qualitätsstufe = MQ200

449393-18L 449393-1L 449393-4L
SIAL449393-18LEA 1330 EUR
SIAL449393-18L SIAL449393-1L SIAL449393-4L
Diethylenglycolmonohexylether, Sigma-Aldrich®
Diethylenglycolmonohexylether
Formel: C₁₀H₂₂O₃
Molecular Weight: 190,28 g/mol
MDL: MFCD00010703
CAS-Nummer: 112-59-4
EINECS: 203-988-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR