3-Picolylbromid Hydrobromid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-(Bromomethyl)pyridine hydrobromide

M-Clarity™ Qualitätsstufe = MQ100

475513-1G 475513-5G
SIAL475513-1GEA 29 EUR
SIAL475513-1G SIAL475513-5G
3-Picolylbromid Hydrobromid, Sigma-Aldrich®
3-Picolylbromid Hydrobromid
Formel: C₆H₇Br₂N
Molecular Weight: 252,94 g/mol
MDL: MFCD01321314
CAS-Nummer: 4916-55-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR