Aluminiumtrifluormethansulfonat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Aluminum trifluoromethanesulphonate, Aluminum triflate, Aluminum tris(trifluoromethanesulphonate)

M-Clarity™ Qualitätsstufe = MQ100

515884-10G 515884-50G
SIAL515884-10GEA 113 EUR
SIAL515884-10G SIAL515884-50G
Aluminiumtrifluormethansulfonat, Sigma-Aldrich®
Aluminiumtrifluormethansulfonat
Formel: C₃AlF₉O₉S₃
Molecular Weight: 474,19 g/mol
MDL: MFCD00143596
CAS-Nummer: 74974-61-1
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR