2-Brom-6-formylpyridin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 6-Bromopyridine-2-carboxaldehyde, 6-Bromopyridine-2-carbaldehyde, 6-Bromo-2-pyridinecarboxaldehyde

M-Clarity™ Qualitätsstufe = MQ100

525693-10G 525693-1G
SIAL525693-10GEA 234 EUR
SIAL525693-10G SIAL525693-1G
2-Brom-6-formylpyridin, Sigma-Aldrich®
2-Brom-6-formylpyridin
Formel: C₆H₄BrNO
Molecular Weight: 186,01 g/mol
MDL: MFCD02683546
CAS-Nummer: 34160-40-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR