4-Chlor-2-iodbenzoesäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

560146-1G 560146-5G
SIAL560146-1GEA 40.6 EUR
SIAL560146-1G SIAL560146-5G
4-Chlor-2-iodbenzoesäure, Sigma-Aldrich®
4-Chloro-2-iodobenzoic acid
Formel: C₇H₄ClIO₂
Molecular Weight: 282,46 g/mol
MDL: MFCD00060676
CAS-Nummer: 13421-13-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR