Chlor(diethyl)phosphin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Diethylchlorophosphine
M-Clarity™ Qualitätsstufe = MQ200
573949-1G
573949-5G
SIAL573949-1GEA
77.9
EUR
SIAL573949-1G
SIAL573949-5G
Chlor(diethyl)phosphin, Sigma-Aldrich®
Chlor(diethyl)phosphin
Formel:
C₄H₁₀ClP Molecular Weight: 124,55 g/mol |
MDL:
MFCD00013621 CAS-Nummer: 686-69-1 EINECS: 211-689-4 UN: 2845 ADR: 4.2,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...