2-Chlor-3-formylpyridin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Chloropyridine-3-carboxaldehyde, 2-Chloronicotinaldehyde, 2-Chloro-3-pyridinecarboxaldehyde

M-Clarity™ Qualitätsstufe = MQ100

632155-1G 632155-5G
SIAL632155-1GEA 26 EUR
SIAL632155-1G SIAL632155-5G
2-Chlor-3-formylpyridin, Sigma-Aldrich®
2-Chlor-3-formylpyridin
Formel: C₆H₄ClNO
Molecular Weight: 141,56 g/mol
MDL: MFCD01315308
CAS-Nummer: 36404-88-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR