3-Brom-4-formylpyridin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Bromo-4-pyridinecarboxaldehyde, 3-Bromoisonicotinaldehyde, 3-Bromopyridine-4-carboxaldehyde

M-Clarity™ Qualitätsstufe = MQ100

638285-1G 638285-5G
SIAL638285-1GEA 64.7 EUR
SIAL638285-1G SIAL638285-5G
3-Brom-4-formylpyridin, Sigma-Aldrich®
3-Brom-4-formylpyridin
Formel: C₆H₄BrNO
Molecular Weight: 186,01 g/mol
MDL: MFCD05864506
CAS-Nummer: 70201-43-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR