1-(4-Chlorbenzyl)piperazin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

650218-1G 650218-5G
SIAL650218-1GEA 72.6 EUR
SIAL650218-1G SIAL650218-5G
1-(4-Chlorbenzyl)piperazin, Sigma-Aldrich®
1-(4-Chlorbenzyl)piperazin
Formel: C₁₁H₁₅ClN₂
Molecular Weight: 210,71 g/mol
MDL: MFCD00040791
CAS-Nummer: 23145-88-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR