1-(4-Brom-2-fluorbenzyl)piperazin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

657719-1G 657719-5G
SIAL657719-1GEA 75.2 EUR
SIAL657719-1G SIAL657719-5G
1-(4-Brom-2-fluorbenzyl)piperazin, Sigma-Aldrich®
1-(4-Brom-2-fluorbenzyl)piperazin
Formel: C₁₁H₁₄BrFN₂
Molecular Weight: 273,15 g/mol
MDL: MFCD07368744
CAS-Nummer: 870703-75-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR