1-(2,6-Dichlorbenzyl)piperazin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

657832-1G 657832-5G
SIAL657832-1GEA 64.6 EUR
SIAL657832-1G SIAL657832-5G
1-(2,6-Dichlorbenzyl)piperazin, Sigma-Aldrich®
1-(2,6-Dichlorbenzyl)piperazin
Formel: C₁₁H₁₄Cl₂N₂
Molecular Weight: 245,15 g/mol
MDL: MFCD02736281
CAS-Nummer: 102292-50-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR