2-Formyl-6-methoxypyridin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
6-Methoxypicolinaldehyde, 6-Methoxypyridine-2-carboxaldehyde, 6-Methoxypyridine-2-carbaldehyde
M-Clarity™ Qualitätsstufe = MQ100
662933-1G
662933-5G
SIAL662933-1GEA
84.9
EUR
SIAL662933-1G
SIAL662933-5G
2-Formyl-6-methoxypyridin, Sigma-Aldrich®
2-Formyl-6-methoxypyridin
Formel:
C₇H₇NO₂ Molecular Weight: 137.14 g/mol |
MDL:
MFCD05664030 CAS-Nummer: 54221-96-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...