Chlor(diphenyl)silan, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

673935-25ML 673935-5ML
SIAL673935-25MLEA 413 EUR
SIAL673935-25ML SIAL673935-5ML
Chlor(diphenyl)silan, Sigma-Aldrich®
Chloro(diphenyl)silane
Formel: C₁₂H₁₁ClSi
Molecular Weight: 218,76 g/mol
MDL: MFCD00045144
CAS-Nummer: 1631-83-0
EINECS: 216-634-8
UN: 2987
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR