Eisen(II)trifluormethansulfonat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Iron (II) Trifluoromethanesulphonate
698172-25G
698172-5G
SIAL698172-25GEA
173
EUR
SIAL698172-25G
SIAL698172-5G
Eisen(II)trifluormethansulfonat, Sigma-Aldrich®
Eisen(II)trifluormethansulfonat
Formel:
C₂F₆FeO₆S₂ Molecular Weight: 353,99 g/mol |
MDL:
MFCD08460524 CAS-Nummer: 59163-91-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...