3-Amino-2-chlor-4-methylpyridin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
3-Amino-2-chloro-4-methylpyridine, 3-Amino-2-chloro-4-picoline, 2-Chloro-4-methylpyridin-3-amine
708135-1G
708135-5G
SIAL708135-1GEA
25
EUR
SIAL708135-1G
SIAL708135-5G
3-Amino-2-chlor-4-methylpyridin, Sigma-Aldrich®
3-Amino-2-chloro-4-methylpyridine
Formel:
C₆H₇ClN₂ Molecular Weight: 142,59 g/mol |
MDL:
MFCD00673152 CAS-Nummer: 133627-45-9 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...