1-Butyl-4-methylpyridiniumiodid, Sigma-Aldrich®
Lieferant: Merck
713112-50G
713112-5G
SIAL713112-50GEA
0
EUR
SIAL713112-50G
SIAL713112-5G
1-Butyl-4-methylpyridiniumiodid, Sigma-Aldrich®
1-Butyl-4-methylpyridinium iodide
Formel:
C₁₀H₁₆IN Molecular Weight: 277,15 g/mol |
CAS-Nummer:
32353-64-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...