3-Furoylchlorid, Sigma-Aldrich®

Lieferant: Merck

715689-1G 715689-5G
SIAL715689-1GEA 61.6 EUR
SIAL715689-1G SIAL715689-5G
3-Furoylchlorid, Sigma-Aldrich®
3-Furoylchlorid
Formel: C₅H₃ClO₂
MDL: MFCD02681970
CAS-Nummer: 26214-65-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR