1-Butyl-4-methylpyridiniumtetrafluoroborat, Sigma-Aldrich®
Lieferant: Merck
73261-50G
73261-5G
SIAL73261-50GEA
256
EUR
SIAL73261-50G
SIAL73261-5G
1-Butyl-4-methylpyridiniumtetrafluoroborat, Sigma-Aldrich®
1-Butyl-4-methylpyridiniumtetrafluoroborate
Formel:
C₁₀H₁₆BF₄N |
MDL:
MFCD03095460 CAS-Nummer: 343952-33-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...