1-(4-(Trifluormethyl)phenyl)piperazin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: N-(α,α,α-Trifluoro-p-tolyl)piperazine

80077-1G-F 80077-5G-F
SIAL80077-1G-FEA 25.9 EUR
SIAL80077-1G-F SIAL80077-5G-F
1-(4-(Trifluormethyl)phenyl)piperazin, Sigma-Aldrich®
1-(4-(Trifluormethyl)phenyl)piperazin
Formel: C₁₁H₁₃F₃N₂
Molecular Weight: 230,23 g/mol
MDL: MFCD00040765
CAS-Nummer: 30459-17-7
EINECS: 250-210-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR