Tetrabutylammoniumtrifluormethansulfonat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

86888-10G 86888-50G
SIAL86888-10GEA 106 EUR
SIAL86888-10G SIAL86888-50G
Tetrabutylammoniumtrifluormethansulfonat, Sigma-Aldrich®
Tetrabutylammonium trifluoromethanesulphonate
Formel: C₁₇H₃₆F₃NO₃S
Molecular Weight: 391,54 g/mol
MDL: MFCD00042585
CAS-Nummer: 35895-70-6
EINECS: 252-783-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR