1-(2-Aminoethyl)piperazin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

A55209-100G A55209-500G
SIALA55209-100GEA 26.4 EUR
SIALA55209-100G SIALA55209-500G
1-(2-Aminoethyl)piperazin, Sigma-Aldrich®
1-(2-Aminoethyl)piperazin
Formel: C₆H₁₅N₃
Molecular Weight: 129,21 g/mol
MDL: MFCD00005971
CAS-Nummer: 140-31-8
EINECS: 205-411-0
UN: 2815
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR