5-Brom-2-furancarbonsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Bromofuran-2-carboxylic acid, 5-Bromofuroic acid

M-Clarity™ Qualitätsstufe = MQ200

B67406-100G B67406-25G
SIALB67406-100GEA 155 EUR
SIALB67406-100G SIALB67406-25G
5-Brom-2-furancarbonsäure, Sigma-Aldrich®
5-Bromo-2-furoic acid
Formel: C₅H₃BrO₃
Molecular Weight: 190,98 g/mol
MDL: MFCD00003239
CAS-Nummer: 585-70-6
EINECS: 209-559-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR