Calciumbis(dihydrogenorthophosphat), Sigma-Aldrich®

Lieferant: Merck

Synonyme: Calcium phosphate, monobasic

C8017-100G C8017-500G
SIALC8017-100GEA 138 EUR
SIALC8017-100G SIALC8017-500G
Calciumbis(dihydrogenorthophosphat), Sigma-Aldrich®
Calciumbis(dihydrogenorthophosphat)
Formel: H₄CaO₈P₂
Molecular Weight: 234,05 g/mol
MDL: MFCD00010898
CAS-Nummer: 7758-23-8
EINECS: 231-837-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR