3-Methylcrotonsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3,3-Dimethylacrylic acid, 3-Methylbut-2-enoic acid

M-Clarity™ Qualitätsstufe = MQ200

D138606-100G D138606-500G
SIALD138606-100GEA 86 EUR
SIALD138606-100G SIALD138606-500G
3-Methylcrotonsäure, Sigma-Aldrich®
3-Methylcrotonsäure
Formel: C₅H₈O₂
Molecular Weight: 100,12 g/mol
MDL: MFCD00004366
CAS-Nummer: 541-47-9
EINECS: 208-782-7
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR