(2-Chlorethyl)diethylammoniumchlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Diethylaminoethylchloride hydrochloride, N-(2-Chloroethyl)-N,N-diethylammonium chloride

M-Clarity™ Qualitätsstufe = MQ200

D87201-100G D87201-1KG
SIALD87201-100GEA 42.4 EUR
SIALD87201-100G SIALD87201-1KG
(2-Chlorethyl)diethylammoniumchlorid, Sigma-Aldrich®
2-Chloroethyldiethylammonium chloride
Formel: C₆H₁₅Cl₂N
Molecular Weight: 172,1 g/mol
MDL: MFCD00012519
CAS-Nummer: 869-24-9
EINECS: 212-786-4
UN: 2928
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR