Nicotinaldehyd, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Pyridine-3-carboxaldehyde, 3-Pyridinecarboxaldehyde, 3-Pyridinecarbaldehyde

M-Clarity™ Qualitätsstufe = MQ200

P62208-100G P62208-25G
SIALP62208-100GEA 101 EUR
SIALP62208-100G SIALP62208-25G
Nicotinaldehyd, Sigma-Aldrich®
Nicotinaldehyd
Formel: C₆H₅NO
Molecular Weight: 107,11 g/mol
MDL: MFCD00006382
CAS-Nummer: 500-22-1
EINECS: 207-900-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR