Isonicotinaldehyd, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Pyridine-4-carboxaldehyde, 4-Pyridinecarboxaldehyde, 4-Pyridinecarbaldehyde

M-Clarity™ Qualitätsstufe = MQ200

P62402-100G P62402-25G P62402-500G
SIALP62402-100GEA 189 EUR
SIALP62402-100G SIALP62402-25G SIALP62402-500G
Isonicotinaldehyd, Sigma-Aldrich®
Isonicotinaldehyd
Formel: C₆H₅NO
Molecular Weight: 107,11 g/mol
MDL: MFCD00006425
CAS-Nummer: 872-85-5
EINECS: 212-832-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR