O15-Palmitoylretinol, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: Retinyl palmitate, Retinol palmitate, alle-trans-Retinylpalmitat

M-Clarity™ Qualitätsstufe = MQ100

R3375-100G R3375-1G
SIALR3375-100GEA 1220 EUR
SIALR3375-100G SIALR3375-1G
O15-Palmitoylretinol, Sigma-Aldrich®
Vitamin A palmitate
Formel: C₃₆H₆₀O₂
Molecular Weight: 524.87 g/mol
MDL: MFCD00019414
CAS-Nummer: 79-81-2
EINECS: 201-228-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR