Vitamin A acetat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: alle-trans-Retinylacetat

M-Clarity™ Qualitätsstufe = MQ200

R7882-1G R7882-5G
SIALR7882-1GEA 69.5 EUR
SIALR7882-1G SIALR7882-5G
Vitamin A acetat, Sigma-Aldrich®
alle-trans-Retinylacetat
Formel: C₂₂H₃₂O₂
Molecular Weight: 328,49 g/mol
MDL: MFCD00019413
CAS-Nummer: 127-47-9
EINECS: 204-844-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR