(-)-Menthon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: L-Menthon

M-Clarity™ Qualitätsstufe = MQ300

W266701-1KG-K W266701-20KG-K W266701-9KG-K
SIALW266701-1KG-KEA 201 EUR
SIALW266701-1KG-K SIALW266701-20KG-K SIALW266701-9KG-K
(-)-Menthon, Sigma-Aldrich®
(-)-Menthon
Formel: C₁₀H₁₈O
Molecular Weight: 154,25 g/mol
MDL: MFCD00001634
CAS-Nummer: 14073-97-3
EINECS: 237-926-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR