Chlor(diphenyl)silan

Lieferant: CHEMPUR SILANE

SILS07725.5 SILS07725.25
SILNSILS07725.5EA 0 EUR
SILNSILS07725.5 SILNSILS07725.25
Chlor(diphenyl)silan
Chloro(diphenyl)silane
Formel: C₁₂H₁₁ClSi
Molecular Weight: 218.76 g/mol
MDL: MFCD00045144
CAS-Nummer: 1631-83-0
EINECS: 216-634-8
UN: 2987
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR