Diphenyldifluorsilan

Lieferant: CHEMPUR SILANE

Synonyme: Difluorodiphenylsilane

SILS07850.25 SILS07850.50
SILNSILS07850.25EA 0 EUR
SILNSILS07850.25 SILNSILS07850.50
Diphenyldifluorsilan
Diphenyldifluorsilan
Formel: C₁₂H₁₀F₂Si
Molecular Weight: 220.29 g/mol
MDL: MFCD00017898
CAS-Nummer: 312-40-3
EINECS: 206-226-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR