Dichlor(ethyl)phenylsilan

Lieferant: CHEMPUR SILANE

SILS08742.1 SILS08742.5 SILS08742.25
SILNSILS08742.1EA 0 EUR
SILNSILS08742.1 SILNSILS08742.5 SILNSILS08742.25
Dichlor(ethyl)phenylsilan
Dichloro(ethyl)(phenyl)silane
Formel: C₈H₁₀Cl₂Si
CAS-Nummer: 1125-27-5
EINECS: 214-407-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR