Chlor(methyl)phenylsilan

Lieferant: CHEMPUR SILANE

SILS11485.5 SILS11485.25
SILNSILS11485.5EA 0 EUR
SILNSILS11485.5 SILNSILS11485.25
Chlor(methyl)phenylsilan
Chloro(methyl)(phenyl)silane
Formel: C₇H₉ClSi
Molecular Weight: 156,69 g/mol
CAS-Nummer: 1631-82-9
EINECS: 216-633-2
UN: 2987
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR