Dichlor(methyl)phenylsilan

Lieferant: CHEMPUR SILANE

Synonyme: Methylphenyldichlorosilane

SILS11500.25 SILS11500.100
SILNSILS11500.25EA 0 EUR
SILNSILS11500.25 SILNSILS11500.100
Dichlor(methyl)phenylsilan
Dichloro(methyl)(phenyl)silane
Formel: C₇H₈Cl₂Si
Molecular Weight: 191,13 g/mol
MDL: MFCD00000490
CAS-Nummer: 149-74-6
EINECS: 205-746-2
UN: 2437
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR