Dichlor(phenyl)silan

Lieferant: CHEMPUR SILANE

SILS13350.5 SILS13350.100
SILNSILS13350.5EA 0 EUR
SILNSILS13350.5 SILNSILS13350.100
Dichlor(phenyl)silan
Dichloro(phenyl)silane
Formel: C₆H₆Cl₂Si
Molecular Weight: 177.1 g/mol
MDL: MFCD00039296
CAS-Nummer: 1631-84-1
EINECS: 216-635-3
UN: 2986
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR