p-Tolyltrimethylsilan
Lieferant: CHEMPUR SILANE
Synonyme:
Trimethyl-p-tolylsilane, Trimethyl(p-tolyl)silane
SILS16675.5
SILS16675.25
SILS16675.100
SILNSILS16675.5EA
0
EUR
SILNSILS16675.5
SILNSILS16675.25
SILNSILS16675.100
p-Tolyltrimethylsilan
p-Tolyltrimethylsilan
Formel:
C₁₀H₁₆Si Molecular Weight: 164,32 g/mol |
MDL:
MFCD00014850 CAS-Nummer: 3728-43-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...