(Brommethyl)cyclopropan

Lieferant: FLUOROCHEM

003713-5G 003713-25G 003713-100G
FLUO003713-5GEA 0 EUR
FLUO003713-5G FLUO003713-25G FLUO003713-100G
(Brommethyl)cyclopropan
(Brommethyl)cyclopropan
Formel: C₄H₇Br
Molecular Weight: 135 g/mol
Siedepunkt: 105 °C (1013 hPa)
Dichte: 1,41 g/cm³ (20 °C)
Lagertemperatur: Kühlschrank
MDL: MFCD00001306
CAS-Nummer: 7051-34-5
EINECS: 230-331-8
UN: 1993
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR