Dimedon
Lieferant: FLUOROCHEM
Synonyme:
Methon
011151-100G
011151-500G
FLUO011151-100GEA
48.8
EUR
FLUO011151-100G
FLUO011151-500G
Dimedon
Dimedon
Formel:
C₈H₁₂O₂ Molecular Weight: 140.18 g/mol Siedepunkt: 234 Schmelzpunkt: 148 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001588 CAS-Nummer: 126-81-8 EINECS: 204-804-4 Merck Index: 13,03268 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...