Dimedon

Lieferant: MENAI

Synonyme: Methon

D1.500 D1.100
MENAD1.500EA 0 EUR
MENAD1.500 MENAD1.100
Dimedon
Dimedon
Formel: C₈H₁₂O₂
Molecular Weight: 140.18 g/mol
Siedepunkt: 234
Schmelzpunkt: 148
Lagertemperatur: Raumtemperatur
MDL: MFCD00001588
CAS-Nummer: 126-81-8
EINECS: 204-804-4
Merck Index: 13,03268

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR