5-Methyl-1,3-cyclohexandion
Lieferant: FLUOROCHEM
|
Achtung
|
Synonyme:
5-Methylcyclohexane-1,3-dione
011195-50G
011195-25G
011195-10G
FLUO011195-50GEA
0
EUR
FLUO011195-50G
FLUO011195-25G
FLUO011195-10G
5-Methyl-1,3-cyclohexandion
5-Methyl-1,3-cyclohexandion
Formel:
C₇H₁₀O₂ Molecular Weight: 126,16 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010379 CAS-Nummer: 4341-24-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...