(Chlormethyl)cyclopropan ≥97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: Cyclopropyl methyl chloride

L06986.06 L06986.14
L06986.06EA 72.7 EUR
L06986.06 L06986.14
(Chlormethyl)cyclopropan ≥97%
(Chlormethyl)cyclopropan

Purity: 97% CAS No.: 5911-08-0 Molecular Formula: C4H7Cl Formula Weight: 90.55 Storage Temperature: 2°C to 8°C Flash Point: -1°(30°F) Refractive Index: 1.4360 Odour: Recognizable Melting Point: -91° Boiling Point: 87-89° Beilstein Reference: 635697 UN No.: 1993 MDL No.: MFCD00001307

Formel: C₄H₇Cl
Molecular Weight: 90,55 g/mol
Siedepunkt: 87…89 °C
Schmelzpunkt: –91 °C
Dichte: 0,980
Flammpunkt: –1 °C (30 °F)
Lagertemperatur: Kühlschrank
MDL: MFCD00001307
CAS-Nummer: 5911-08-0
EINECS: 227-632-1
UN: 1993
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR