Cyclopropanmethylamin ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: (Aminomethyl)cyclopropan, (Cyclopropylmethyl)amin

A13946.03 A13946.06 A13946.14
A13946.03EA 29.1 EUR
A13946.03 A13946.06 A13946.14
Cyclopropanmethylamin ≥98%
Cyclopropanmethylamin

Purity: 98+% CAS No.: 2516-47-4 Molecular Formula: C4H9N Formula Weight: 71.12 Flash Point: 2°(36°F) Refractive Index: 1.4330 Odour: Amine-like Boiling Point: 85-86° Beilstein Reference: 773674 UN No.: 2733 MDL No.: MFCD00037147

Formel: C₄H₉N
Molecular Weight: 71,12 g/mol
Siedepunkt: 85…86 °C
Dichte: 0,820
Flammpunkt: 2 °C (36 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00037147
CAS-Nummer: 2516-47-4
EINECS: 219-737-6
UN: 2733
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR