Cyclopenten 98

Lieferant: Thermo Fisher Scientific
Gefahr

41461.AC 41461.AE
41461.ACEA 40.2 EUR
41461.AC 41461.AE
Cyclopenten 98
Cyclopentene

Purity: 98+% CAS No.: 142-29-0 Molecular Formula: C5H8 Formula Weight: 68.12 Flash Point: -29°(-20°F) Physical Form: Liquid Density: 0.773 Refractive Index: 1.4225 Sensitivity: Air Sensitive Appearance: Colourless Melting Point: -135° Boiling Point: 44-45° Beilstein Reference: 635707 UN No.: 2246 MDL No.: MFCD00001394

Formel: C₅H₈
Molecular Weight: 68.12 g/mol
Siedepunkt: 44…45 °C
Schmelzpunkt: –135 °C
Dichte: 0.773
Flammpunkt: –29 °C (–20 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00001394
CAS-Nummer: 142-29-0
EINECS: 205-532-9
UN: 2246
ADR: 3 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR