2,7-Dibromfluoren

Lieferant: FLUOROCHEM
Achtung

Synonyme: 2,7-Dibromo-9H-fluorene

005333-25G 005333-5G
FLUO005333-25GEA 121 EUR
FLUO005333-25G FLUO005333-5G
2,7-Dibromfluoren
2,7-Dibromfluoren
Formel: C₁₃H₈Br₂
Molecular Weight: 324,01 g/mol
MDL: MFCD00019048
CAS-Nummer: 16433-88-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR