tri-Lithiumhexafluoraluminat

Lieferant: Honeywell Chemicals

01156.9030 01156.91200
HONE01156.9030EA 0 EUR
HONE01156.9030 HONE01156.91200
tri-Lithiumhexafluoraluminat
tri-Lithiumhexafluoraluminat
Formel: AlF₆Li₃
Molecular Weight: 161,79 g/mol
CAS-Nummer: 13821-20-0
EINECS: 237-509-4
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR