Natriumhexafluoroaluminat ≥99,5% (Metall-Basis)
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Trinatriumhexafluoraluminat
36345.18
36345.30
36345.18EA
125
EUR
36345.18
36345.30
Natriumhexafluoroaluminat ≥99,5% (Metall-Basis)
Natriumhexafluoroaluminat
Purity: 99.5%
CAS No.: 13775-53-6
Molecular Formula: Na3AlF6
Formula Weight: 209.95
Physical Form: 3-12mm Lump
Density: 2.90
Melting Point: 1000°
UN No.: 3260
MDL No.: MFCD00003507
Formel:
AlF₆Na₃ Molecular Weight: 209,94 g/mol Schmelzpunkt: 1000 °C Dichte: 2,90 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003507 CAS-Nummer: 13775-53-6 EINECS: 237-410-6 UN: 3077 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...